This item is now discontinued. Please browse similar items

Labconco 9907200 Vacuum Pump Storage Cabinet with Single Right-Hinged Door for 24" Fume Hood

Representative image only.
Labconco Mfr # 9907200 – Item # WZ-09115-59
Provide support for your fume hood while adding additional storage
  • Choose between four types of storage cabinets – Acid, Solvent, Standard, and Vacuum Pump
  • Constructed of powder-coated steel
  • Supports loads of up to 800 pounds
  • Designed with a signle right-hinged door
  • Suitable for 24" fume hoods
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Hood Width (in)24
  • Exterior Width (in)24
  • Exterior Height (in)36.75
  • Exterior Depth (in)22
  • Shipping Weight (lbs)110
  • DescriptionVacuum Pump Storage Cabinet with Single Right-Hinged Door for 24\" Fume Hood
  • Warranty1 Year
1 Year Warranty

Key Features

  • Choose between four types of storage cabinets – Acid, Solvent, Standard, and Vacuum Pump
  • Constructed of powder-coated steel
  • Supports loads of up to 800 pounds
  • Designed with a signle right-hinged door
  • Suitable for 24" fume hoods
  • 1 year warranty

More About this Item

Standard base cabinets are ideal for general storage of small laboratory equipment, mild acids ,and other reagents.

Acid storage cabinets allow you to safely store and vent acids and other corrosive liquids. The interior is constructed with a corrosion-resistant polyethylene liner to enhance corrosion resistance.

Solvent storage cabinets are suitable for storing and venting solvents and other flammable liquids. These cabinets will protect the interior contents in case of an external fire. These cabinets are constructed with dual wall powder-coated steel and feature a 1.5” air gap for fire protection.

Vacuum pump storage cabinets are utilized for vacuum pump storage.

Work surfaces are sold separately and required. Spillstopper work surfaces keep accidental spills and liquids away from the seam between the hood wall and surrounding countertop.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat