Lab-Crest 110-700 Coupling Component, male part, SS, for use with 3 oz reaction vessel only
Representative image only.
Lab-Crest Mfr # 110-700-001 – Item # WZ-63400-76
£177.25GBP / Each
£212.70GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Materialstainless steel
  • Compatible WithLab-Crest Pressure Reaction Vessels
  • DescriptionCoupling Component, male part, SS, for use with 3 oz reaction vessel only
More About this Item
Parts and accessories for your Lab-Crest system.
A complete coupling consists of a female coupling (63400-64 or 63400-72), male coupling (63400-66), insert (63400-68 or 63400-78), and an o-ring.