This item is now discontinued. Please browse similar items
Kruss PRG-100-ETT SS Polarimeter Tube, temperature-controllable with temperature sensor, 100-mm
Representative image only.
Kruss Mfr # PRG-100-ETT – Item # WZ-81250-45
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSS Polarimeter Tube, temperature-controllable with temperature sensor, 100-mm
  • Warranty2 Years
Two year warranty
Key Features
  • 2 year warranty
More About this Item
Plates and tubes customize your polarimeter to your application.
Optional stainless steel tubes with temperature sensor allow you to directly monitor the sample temperature and compensate your readings.