This item is now discontinued. We recommend the following as suggested replacements: WZ-14669-05
Koflo 3/8-12PPT Mounted in-line mixer; 3/8" tube OD, 12 elements, 10/pack
Representative image only.
Koflo Mfr # 3/8-12PPT – Item # WZ-04668-04
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolyacetal
  • No. Of Elements12
  • Tubing OD (in)0.375
  • Tubing OD (mm)9.53
  • Mixer Length (in)4.25
  • Mounting Type0.375\" plain end
  • DescriptionMounted in-line mixer; 3/8\" tube OD, 12 elements, 10/pack
  • WarrantyLimited Lifetime
Limited Lifetime Warranty
Key Features
  • Limited Lifetime Warranty
More About this Item
Mounted mixers can be used with any compression fitting with the same tubing OD.