This item is now discontinued. Please browse similar items
Koflo 2-SA-3-12R-11 Sanitary Mixer, 2" Tri-clamp size, 12 elements
Representative image only.
Koflo Mfr # 2-SA-3-12R-11 – Item # WZ-04668-46
  • Limited Lifetime Warranty
SPECS & DESCRIPTION
Specifications & Description
  • Material316 Stainless Steel
  • No. Of Elements12
  • Pipe Id (in)1.87
  • Pipe Id (mm)47.5
  • Mixer Length (in)30
  • Mounting Type2\" Tri-Clamp
  • DescriptionSanitary Mixer, 2\" Tri-clamp size, 12 elements
  • WarrantyLTD Life Time
Limited Lifetime Warranty
Key Features
  • Limited Lifetime Warranty
More About this Item
Blade Design Mixers are best for low-viscosity (<1,500 Cp) fluids. The faster the fluid moves, the faster (and more thorough) the mixing. Great for mixing products into water or water-like fluids.