This item is now discontinued. Please browse similar items
Koflo 2.5-SA-3-12RH-11 Sanitary Mixer, helical design, 2-1/2" Tri-clamp size, 12 elements
Representative image only.
Koflo Mfr # 2.5-SA-3-12RH-11 – Item # WZ-04668-86
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Material316 Stainless Steel
  • No. Of Elements12
  • Pipe Id (in)1.87
  • Pipe Id (mm)47.5
  • Mixer Length (in)44
  • Mounting Type2 1/2\" Tri-Clamp
  • DescriptionSanitary Mixer, helical design, 2-1/2\" Tri-clamp size, 6 elements
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Helical Design Mixers are best for high-viscosity (>1,500 Cp) fluids, and when fluids are moving more slowly through the mixer. Excellent for combining products into fluids such as yogurt, mayonnaise, and ice cream.