This item is now discontinued. Please browse similar items
Koflo 1-SA-3-12RH-11 Sanitary Mixer, helical design, 1" Tri-clamp size, 12 elements
Representative image only.
Koflo Mfr # 1-SA-3-12RH-11 – Item # WZ-04668-80
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Material316 Stainless Steel
  • No. Of Elements12
  • Pipe Id (in)0.87
  • Pipe Id (mm)22.1
  • Mixer Length (in)18
  • Mounting Type1\" Tri-Clamp
  • DescriptionSanitary Mixer, helical design, 1\" Tri-clamp size, 12 elements
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Helical Design Mixers are best for high-viscosity (>1,500 Cp) fluids, and when fluids are moving more slowly through the mixer. Excellent for combining products into fluids such as yogurt, mayonnaise, and ice cream.