Koehler K61109 Centrifuge Tube, Cone, Capillary Tip; 100 mL
Representative image only.
Koehler Mfr # K61109 – Item # WZ-17300-33
£108.35GBP / Each
£130.02GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Tube Sizes Accepted100 mL, capillary tip capable of measuring 0.01 mL and readable by estimation to 0.005%; cone-shaped
  • Max Tube Length (in)8
  • DescriptionCentrifuge Tube, Cone, Capillary Tip; 100 mL
More About this Item
Parts and accessories to meet your application needs, and keep your Koehler Centrifuge operating smoothly.