Koehler K46001 Cloud and Pour Point Chamber with Inlet and Outlet Fittings
Representative image only.
Koehler Mfr # K46001 – Item # WZ-59861-02
Indicators of the lowest temperature of utility for petroleum products
  • Conforms to ASTM D 97, D 2500, and related specifications
£2,182.10GBP / Each
£2,618.52GBP (incl VAT)
Cole-Parmer Clean-Wipes, 70% Alcohol/30% DI Water; 100/Canister
£8.50GBP / Pkg of 100
£10.20GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • ASTM MethodD 97/D 2500
  • Height (in)12
  • Height (cm)30.48
  • DescriptionCloud and Pour Point Chamber with Inlet and Outlet Fittings
Key Features
  • Conforms to ASTM D 97, D 2500, and related specifications
More About this Item
Cloud and Pour Point Chamber allows you to immerse four test jackets in suitable freezing mixtures. Top plate and 1/2" cork insulation aid in cold retention. Includes copper jackets, gaskets, disks, and thermometer holders for test jars. Test jars and thermometers sold separately. Measures 10-1/2" dia x 12"H.
Cloud and Pour Point Bath features three jacket refrigerated baths, each preset to a different temperature: -30, 0, and 30°F (35, 18, and 1°C). Synthetic sponge covers top plate; gasketed hoods reduce ice accumulation. Includes copper jackets, gaskets, disks, and thermometer holders for test jars. Stainless steel cabinet and interior. Measures 24"L x 18"W x 23"H.