This item is now discontinued. Please browse similar items

KNF UN026ANI EX Explosion Proof Motor Vacuum Pump, Al/NBR; 0.9 cfm/26.9"Hg-36psi

Representative image only.
KNF Mfr # 072942/075636 – Item # WZ-07057-84
Oversized, permanently lubricated bearings for long-term operation
  • Ideal for hazardous areas where flammable gases or vapors may be present
  • Rated for continuous, heavy-duty use
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Free-Air Capacity (CFM)0.9
  • Free-Air Capacity (LPM)25.5
  • Free-Air Capacity (LPH)1530
  • Max Vacuum (in Hg)26.9
  • Max Vacuum (mbar)102.4
  • Max Vacuum (mm Hg)76.7
  • Max Vacuum (torr)76.7
  • Port Size1/8\" NPT(F)
  • Max Temperature (° F)105
  • Max Temperature (° C)40
  • Duty CycleContinuous
  • Power (VAC)115 / 230
  • Power (amps)2.2
  • Power (Hz)50/60
  • Height (in)10.9375
  • Width (in)5.8125
  • Length (in)12.9375
  • Height (cm)27.78
  • Width (cm)14.76
  • Length (cm)32.86
  • Height (mm)277.8
  • Width (mm)147.6
  • Length (mm)328.6
  • DescriptionExplosion Proof Motor Vacuum Pump, Al/NBR; 0.9 cfm/26.9\"Hg-36psi
  • Warranty1 Year
1 Year Warranty

Key Features

  • Ideal for hazardous areas where flammable gases or vapors may be present
  • Rated for continuous, heavy-duty use
  • 1 year warranty

More About this Item

Diaphragm pumps are fitted with Class I, Division 1, Groups C & D explosion-proof motors. Designed for continuous, heavy-duty use, ball bearings are used throughout to provide exceptional reliability. Models are fitted with a pre-wired connection box and are tested and ready to install into your system.

Contact one of our Application Specialists for assistance with selecting the proper pump for your needs.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat