This item is now discontinued. Please browse similar items
Klarmann Rulings KR-613-22.8MM Reticle, Cross Scale Pattern, 22.8 mm dia
Representative image only.
Klarmann Rulings Mfr # KR-613-22.8MM – Item # WZ-46800-25
Add a scale to your microscope inspections for increased accuracy and precision
SPECS & DESCRIPTION
Specifications & Description
  • Eyepiece Diameter (mm)22.8
  • DescriptionReticle, Cross Scale Pattern, 22.8 mm dia
More About this Item
Designed to increase the accuracy of your inspections, these clear glass discs are marked with precise scales or patterns. Reticles fit into microscope eyepieces at the intermediate image area of the specimen or object being examined. Both the scale or pattern of the reticle and the object image are superimposed and in focus.