This item is now discontinued. Please browse similar items
Kinetic active-air Vibration-Free Platform, Feromagnetic Stainless 20" x 24" Active-Air
Representative image only.
Kinetic Mfr # 2214-01-12 – Item # WZ-01019-47
Enhance the performance of your balances with a vibration-free tabletop
  • Low profile passive- or active-air designs
  • Excellent horizontal isolation
  • ISO Class 5 (formerly Class 100) cleanroom compatible
  • 6 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Width (in)24
  • Depth (in)20
  • DescriptionVibration-Free Platform, Feromagnetic Stainless 20\" x 24\" Active-Air
  • Warranty6 month warranty
6 Month Warranty
Key Features
  • Low profile passive- or active-air designs
  • Excellent horizontal isolation
  • ISO Class 5 (formerly Class 100) cleanroom compatible
  • 6 month warranty
More About this Item
These platforms reduce vibration and shock when using precision testing equipment, such as balances. There are two configurations, the passive-air and active-air platforms. The passive-air design requires an optional hand pump if an air source is not available. It requires periodic manual leveling. The active-air design requires an air source (not included) for use. It is a self-leveling unit and recommended for regular use. The load capacity on all models is 200 lb.
Model 01019-47 is the enhanced version of all of the platforms: It incorporates multistage horizontal isolation for applications where horizontal vibrations are a problem.