Cole-Parmer TELOS® Top Frit for Dry Loading Cartridge, Luer Lock, 40 g, 20 µm; 50/pk

Representative image only.
Cole-Parmer Item # WZ-06475-39
Extend the life of your dry loading cartridges with these replacement parts
  • Constructed of durable polyethylene
  • Compatible with TELOS 40 g dry loading cartridge with luer lock
  • 20 µ fritted disk

PRODUCT OPTIONS

£138.45GBP / Pkg of 50
£166.14GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • MaterialPolyethylene
  • Compatible WithColumn 06475-59; 20 µm
  • DescriptionTop Frit for Dry Loading Cartridge, Luer Lock, 40 g, 20 µm; 50/pk

Key Features

  • Constructed of durable polyethylene
  • Compatible with TELOS 40 g dry loading cartridge with luer lock
  • 20 µ fritted disk

More About this Item

Choose from a variety of replacement top frit sizes for your Cole-Parmer TELOS dry loading columns.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat