This item is now discontinued. Please browse similar items
Cole-Parmer TELOS® Specialty SPE Column, PSA, 100 mg Sorbent, 1 mL; 100/Pk
Representative image only.
Cole-Parmer Item # WZ-06476-26
Ideal for extraction of strong acids and polyacidic compounds from aqueous samples
  • Manufactured with PSA (Primary Secondary Amine) solvent
SPECS & DESCRIPTION
Specifications & Description
  • Bed Mass (mg)100
  • Total Volume Capacity (mL)1
  • SorbentPSA
Key Features
  • Manufactured with PSA (Primary Secondary Amine) solvent
More About this Item
PSA (Primary Secondary Amine) is a weak anion exchange SPE sorbent used to extract strong acids and polyacidic compounds from aqueous samples. The weak anion exchange sorbent allows analytes to be eluted by eliminating the charge on the sorbent at high pH. The sorbent is supplied in the free base form. Columns are made of polypropylene and feature Luer-slip outlet.