This item is now discontinued. Please browse similar items
Cole-Parmer TELOS® Specialty SPE Column, Neutral Alumina (AL-N), 500 mg Sorbent, 6 mL; 30/Pk
Representative image only.
Cole-Parmer Item # WZ-06476-46
Easily isolate compounds from nonpolar solvents
  • Polar SPE sorbents
SPECS & DESCRIPTION
Specifications & Description
  • Bed Mass (mg)500
  • Total Volume Capacity (mL)6
  • SorbentAlumina—neutral (AL-N)
Key Features
  • Polar SPE sorbents
More About this Item
Alumina (acidic, basic, and neutral) are polar SPE sorbents commonly used to isolate compounds from nonpolar solvents. Columns are made of polypropylene and feature Luer-slip outlet.