This item is now discontinued. Please browse similar items
Cole-Parmer TELOS® Specialty SPE Column, Cation/Nonpolar (H-CX), 500 mg Sorbent, 3 mL; 50/Pk
Representative image only.
Cole-Parmer Item # WZ-06477-85
Optimized for basic drug extraction from biological fluids
  • Manufactured with H-CX (hydrophobic/cation) sorbent
SPECS & DESCRIPTION
Specifications & Description
  • Bed Mass (mg)500
  • Total Volume Capacity (mL)3
  • SorbentH-CX (hydrophobic cation)
Key Features
  • Manufactured with H-CX (hydrophobic/cation) sorbent
More About this Item
H-CX (hydrophobic/cation) is a mixed-mode sorbent optimized for basic drug extraction from biological fluids. The dual retention mechanism allows an extensive interference elution regime, removing salts, polar interferences, peptides, and proteins prior to analyte elution. H-CX is a commonly use sorbent for many drugs of abuse applications. Columns are made of polypropylene and feature Luer-slip outlet.