This item is now discontinued. Please browse similar items
Cole-Parmer TELOS® QuEChERS Extraction Kit w/o Tube, 6 g MgSO4, 1.5 g NaCl; 50/pk
Representative image only.
Cole-Parmer Item # WZ-06478-86
Quickly and economically extract the required analytes into solution
  • Salt pack helps transfer salt to the centrifuge tube quickly and easily with minimal waste
  • Conform with documented AOAC 2007.01 and EN 15662 protocols
  • Break up complex sample types with homogenizer balls
SPECS & DESCRIPTION
Specifications & Description
  • Total Volume Capacity (mL)50
  • Salts6 g MgSO4 (anhydrous), 1.5 g NaCl
  • MethodologyNone
  • DescriptionKit without tubes
Key Features
  • Salt pack helps transfer salt to the centrifuge tube quickly and easily with minimal waste
  • Conform with documented AOAC 2007.01 and EN 15662 protocols
  • Break up complex sample types with homogenizer balls
More About this Item
TELOS QuEChERS extraction kits offer the first stage cleanup for many sample types. This initial crude extraction and partitioning step uses organic solvent and salt solution to extract the required analytes into solution. Select from extraction kits without a tube, kits with a 50 mL centrifuge tube, or kits with a tube and homogenizer beads. Beads aid in breaking up more complex sample types.