Cole-Parmer TELOS® PPT Protein Precipitation Columns, 1 mL; 100/PK
Representative image only.
Cole-Parmer Item # WZ-06482-37
Efficient and reliable protein precipitation by filtration
  • Ideal for small and varying sample numbers
£199.02GBP / Pkg of 100
£238.82GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Media Volume (mL)1
  • Fritted Disk2
  • ApplicationsProtein precipitation
  • DescriptionTELOS® PPT protein precipitation columns; 100/PK
Key Features
  • Ideal for small and varying sample numbers
More About this Item
Clean up biological fluid samples, especially plasma, using TELOS PPT protein precipitation columns. Complete protein precipitation process takes place in the column—no offline liquid handling and centrifugation steps are required. Specially designed double frits provide fast and trouble-free protein precipitation. One frit holds up the precipitation solvent, acetonitrile, while the other frit with larger porosity prevents blocking, allowing reproducible collection of the subsequent filtrate.
Dispense sample directly into the acetonitrile with force to achieve reproducible precipitation from sample to sample. The optimum ratio of acetonitrile to plasma is 4:1 v/v. This ensures complete precipitation occurs and the filtrate is precipitate free.