Cole-Parmer TELOS® Polar SPE Column, silica, 200 mg sorbent, 3 mL; 50/pk
Representative image only.
Cole-Parmer Item # WZ-06476-08
Extract polar analytes from nonpolar organic solvents
  • Compatible with manual and automated sample protocols
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • Bed Mass (mg)200
  • Column Particle Size (µm)50
  • Wetted MaterialsPolypropylene body and polyethylene frits
  • Total Volume Capacity (mL)3
  • SorbentSilica
Key Features
  • Compatible with manual and automated sample protocols
More About this Item
Use TELOS polar (normal phase) silica SPE columns to retain the analytes of interest or to remove polar interferences from the sample. Silica sorbent has a pore diameter of 70Å. All columns are made of polypropylene and feature Luer-slip outlet.