Cole-Parmer TELOS® Phase Separator, 150 mL; 25/pk
Representative image only.
Cole-Parmer Item # WZ-06475-52
Quickly and easily separate aqueous and chlorinated solvents
  • Gravity flow filtration process, no vacuum required
  • No need for separating funnels and rewashing glassware
PRODUCT OPTIONS
£124.54GBP / Pkg of 25
£149.45GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Total Volume Capacity (mL)150
Key Features
  • Gravity flow filtration process, no vacuum required
  • No need for separating funnels and rewashing glassware
More About this Item
Simply apply aqueous and chlorinated solvents to the column for a quick separation. A special hydrophobic frit allows the chlorinated solvent to pass through under gravity while preventing the aqueous layer from flowing though. The aqueous layer remains in the separator column, separating the two phases and allowing additional work up.