This item is now discontinued. Please browse similar items
Cole-Parmer TELOS® Nonpolar SPE Microplate, C18, 10 mg sorbent, 96 fixed wells; 1/pk
Representative image only.
Cole-Parmer Item # WZ-06476-57
Optimized for sample processing and extraction of small volume biological fluids
  • Select from three main retention mechanisms: nonpolar, ion exchange, or polar
  • Use with vacuum or positive pressure
  • Applications: Nonpolar
SPECS & DESCRIPTION
Specifications & Description
  • Bed Mass (mg)10
  • Carbon Loading17.5%
  • Column Particle Size (µm)50
  • End CapNo
  • ApplicationsNonpolar
  • SorbentC18
Key Features
  • Select from three main retention mechanisms: nonpolar, ion exchange, or polar
  • Use with vacuum or positive pressure
  • Applications: Nonpolar
More About this Item
The internal diameter, frit design, and sorbent material of the Cole-Parmer® TELOS® Silica-Based SPE fixed 96-well microplates allow for maximum recovery with minimal sample dilution and reduced evaporation time. Select endcapped sorbents for applications requiring minimal secondary silanol interactions to enhance compound retention and elution. C4 (WP) sorbents with average pore size of 300 Å are ideal for compounds with molecular weight >10,000.