Cole-Parmer TELOS® neo™ WAX MicroPlate™ SPE Microplate, populated, 10 mg sorbent; 1/ea
Representative image only.
Cole-Parmer Item # WZ-06474-03
50 µL elution volume for minimal sample dilution and reduced evaporation time
  • Versatile 96-well format for small volume biological fluid samples
  • Optimized well geometry for 5 and 10 mg sorbent masses
  • Use with vacuum or positive pressure
  • Applications: Mixed-mode weak anion exchange sorbent for strong acidic analyte extraction
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • Bed Mass (mg)10
  • Column Particle Size (µm)60
  • Bottom TypePolymer based
  • ApplicationsMixed-mode weak anion exchange
  • SorbentWAX
  • DescriptionWAX MicroPlate SPE Microplate, populated, 10 mg sorbent; 1/ea
Key Features
  • Versatile 96-well format for small volume biological fluid samples
  • Optimized well geometry for 5 and 10 mg sorbent masses
  • Use with vacuum or positive pressure
  • Applications: Mixed-mode weak anion exchange sorbent for strong acidic analyte extraction
More About this Item
The internal diameter, frit design, and sorbent material of the Cole-Parmer® TELOS® neo™ MicroPlate™ 96-well plates allow for maximum recovery with minimal sample dilution and reduced evaporation time. A well packed with 5 mg sorbent allows analytes to be eluted in as little as 50 µL.
Populated plate comes with 96 wells already in place. Modular design lets you simply remove any unwanted wells and add sealing strips. Full or partially populated plates can be processed using either vacuum or positive pressure.