This item is now discontinued. Please browse similar items
Cole-Parmer TELOS® neo™ WAX MicroPlate™ SPE Microplate, loose wells, 10 mg sorbent; 100/pk
Representative image only.
Cole-Parmer Item # WZ-06474-08
Populate base plate with only the wells needed
  • 50 µL elution volume for minimal sample dilution and reduced evaporation time
  • Optimized well geometry for 5 and 10 mg sorbent masses
  • Use with vacuum or positive pressure
  • Applications: Mixed-mode weak anion exchange sorbent for strong acidic analyte extraction
SPECS & DESCRIPTION
Specifications & Description
  • Bed Mass (mg)10
  • Column Particle Size (µm)60
  • Bottom TypePolymer based
  • ApplicationsMixed-mode weak anion exchange
  • SorbentWAX
  • DescriptionWAX MicroPlate SPE Microplate, loose wells, 10 mg sorbent; 100/pk
Key Features
  • 50 µL elution volume for minimal sample dilution and reduced evaporation time
  • Optimized well geometry for 5 and 10 mg sorbent masses
  • Use with vacuum or positive pressure
  • Applications: Mixed-mode weak anion exchange sorbent for strong acidic analyte extraction
More About this Item
The internal diameter, frit design, and sorbent material of the Cole-Parmer® TELOS® neo™ MicroPlate™ SPE loose wells allow for maximum recovery with minimal sample dilution and reduced evaporation time. A well packed with 5 mg sorbent allows analytes to be eluted in as little as 50 µL.
Use the loose wells for assays when the sample numbers vary, populating the base plate with only the required number of wells. Base plates are required to use loose wells. Full or partially populated plates can be processed using either vacuum or positive pressure. Order method development kit to build a method development plate to suit your application.
Use the loose-well plates for assays when the sample numbers vary, populating the base plate with only the required number of wells. Base plates are required to use loose-well plates. Order method development kit to build a method development plate to suit your applicatiion.