This item is now discontinued. Please browse similar items
Kinesis TELOS® neo™ WAX Fixed-Well Plate, 10 mg sorbent; 1/ea
Representative image only.
Cole-Parmer Item # WZ-06474-40
Modular design lets you simply remove any unwanted wells and add sealing strips
  • Quickly obtain clean extracts and reduce time spent on method development
  • Faster method development saves you money
  • Water-wettable polymer is not affected by drying out
  • Applications: Mixed-mode weak anion exchange
SPECS & DESCRIPTION
Specifications & Description
  • Bed Mass (mg)10
  • Column Particle Size (µm)60
  • Bottom TypePolymer based
  • ApplicationsMixed-mode weak anion exchange
  • SorbentWAX
  • DescriptionWAX Fixed-Well Plate, 10 mg sorbent; 1/ea
Key Features
  • Quickly obtain clean extracts and reduce time spent on method development
  • Faster method development saves you money
  • Water-wettable polymer is not affected by drying out
  • Applications: Mixed-mode weak anion exchange
More About this Item
Cole-Parmer® TELOS® neo™ populated plates come with 96 wells already in place—ideal for high throughput assays. Polymer-based sorbents have no silanol interactions, excellent flow characteristics, and pH stability providing highly reproducible results. Elution volumes are kept to a minimum by allowing maximum interaction between the analyte and sorbent.