Cole-Parmer TELOS® Multilayer SPE Column, PSA / C18, 500 mg sorbent, 6 mL; 30/pk
Representative image only.
Cole-Parmer Item # WZ-06477-89
Achieve superior clean up when conducting multiresidue analysis
  • Save money by extracting multiple suites with a single column
  • Clean up of extracts from fruits and vegetable samples prior to analysis of pesticides and herbicides
PRODUCT OPTIONS
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • Bed Mass (mg)500
  • End CapNo
  • Total Volume Capacity (mL)6
  • SorbentPSA/C18
Includes
Polyethylene frits
Key Features
  • Save money by extracting multiple suites with a single column
  • Clean up of extracts from fruits and vegetable samples prior to analysis of pesticides and herbicides
More About this Item
Multilayered SPE columns are designed for efficient extraction of multi-analyte suites. Layers of different SPE sorbents in a single SPE column allow a broader range of analytes to be extracted than is possible with a single sorbent. Each column features 20 µm frits at the top, bottom, and between sorbent layers. Columns feature Luer-slip outlet.
Columns are made of polypropylene.