This item is now discontinued. Please browse similar items
Cole-Parmer TELOS® Multilayer SPE Column, Na2SO4 / MgO3Si, 4 g sorbent, 15 mL; 20/pk
Representative image only.
Cole-Parmer Item # WZ-06476-37
Achieve superior clean up when conducting multiresidue analysis
  • Save money by extracting multiple suites with a single column
  • Remove water from water immiscible solvents
SPECS & DESCRIPTION
Specifications & Description
  • Bed Mass (mg)4000
  • End CapNo
  • Total Volume Capacity (mL)15
  • SorbentNa₂SO₄/MgO₃Si
Includes
Polyethylene frits
Key Features
  • Save money by extracting multiple suites with a single column
  • Remove water from water immiscible solvents
More About this Item
Multilayered SPE columns are designed for efficient extraction of multi-analyte suites. Layers of different SPE sorbents in a single SPE column allow a broader range of analytes to be extracted than is possible with a single sorbent. Each column features 20 µm frits at the top, bottom, and between sorbent layers. Columns feature Luer-slip outlet.
Columns are made of polypropylene.