This item is now discontinued. Please browse similar items
Cole-Parmer TELOS® Multilayer SPE Column, Carbon / PSA / SAX, 1.5 g sorbent, 15 mL; 20/pk
Representative image only.
Cole-Parmer Item # WZ-06477-88
Achieve superior clean up when conducting multiresidue analysis
  • Save money by extracting multiple suites with a single column
  • Clean up of extracts from fruits and vegetable samples prior to analysis of pesticides and herbicides
SPECS & DESCRIPTION
Specifications & Description
  • Bed Mass (mg)1500
  • End CapYes
  • Total Volume Capacity (mL)15
  • SorbentCarbon/PSA/SAX
Includes
Polyethylene frits
Key Features
  • Save money by extracting multiple suites with a single column
  • Clean up of extracts from fruits and vegetable samples prior to analysis of pesticides and herbicides
More About this Item
Multilayered SPE columns are designed for efficient extraction of multi-analyte suites. Layers of different SPE sorbents in a single SPE column allow a broader range of analytes to be extracted than is possible with a single sorbent. Each column features 20 µm frits at the top, bottom, and between sorbent layers. Columns feature Luer-slip outlet.
Columns are made of polypropylene.