This item is now discontinued. Please browse similar items

Kinesis TELOS® HPLC Column, C18 AB-D, 5 µm, 150 mm L x 4.6 mm ID

Representative image only.
Kinesis Item # WZ-06480-18
Chromatography columns in a comprehensive array of high-density bonded phases
  • Supported by 1-cm guard cartridges with identical packing material Includes test chromatogram
  • C18 AB-D phase
  • Suitable for HPLC systems

SPECS & DESCRIPTION

Specifications & Description

  • PhaseC18 AB-D
  • Column Length (cm)15
  • Column Length (mm)150
  • Column Inside Diameter (ID) (mm)4.6
  • Column Particle Size (µm)5
  • Max Pressure (PSI)10000
  • Max Operating Temperature (° C)60
  • DescriptionHPLC Column, C18 AB-D, 5 µm, 150 mm L x 4.6 mm ID

Key Features

  • Supported by 1-cm guard cartridges with identical packing material Includes test chromatogram
  • C18 AB-D phase
  • Suitable for HPLC systems

More About this Item

Kinesis® TELOS® columns deliver better performance, reliability, and reproducibility. Each is individually tested for quality and includes a test chromatogram, which lists the test conditions and analytes.

Specially deactivated to produce superior peak shapes and performance for amines and acids. TELOS ultra pure silica selection, proprietary bonding, endcapping reagents, and procedures yield columns that are much more resistant to degradation by acidic and basic mobile phase compositions, while improving peak shapes for amines and acids as a result of the reduced negative surface interactions.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat