Cole-Parmer TELOS® Flash Chromatography Column, Luer lock, Silica, 40 g; 10/pk
Representative image only.
Cole-Parmer Item # WZ-06474-76
Use with SP and Idolera™ systems from Biotage®
  • Optimized column geometry provides maximum recovery
  • Excellent for purification of a wide range of synthetic and natural compounds
PRODUCT OPTIONS
£117.99GBP / Pkg of 10
£141.59GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Bed Mass (g)40
  • Column Particle Size (µm)50
  • Wetted MaterialsPolypropylene column, polyethylene frit
  • Max Pressure (PSI)180
  • SorbentSilica
Key Features
  • Optimized column geometry provides maximum recovery
  • Excellent for purification of a wide range of synthetic and natural compounds
More About this Item
Make connections to flash chromatography instruments quick, easy, and secure with Luer-lok connectors. Columns are made of polypropylene and have pressure ratings to support common flash chromatography back pressures.