This item is now discontinued. Please browse similar items
Kinesis TELOS® Endcapped Nonpolar SPE Column, C8, 200 mg sorbent, 3 mL; 50/pk
Representative image only.
Kinesis Item # WZ-06477-09
Extract hydrophobic or polar organic analytes from aqueous matrices
  • Features C8 sorbent
  • Applications: Extraction of hydrophobic and polar analytes
SPECS & DESCRIPTION
Specifications & Description
  • Bed Mass (mg)200
  • Carbon Loading9%
  • Column Particle Size (µm)50
  • End CapYes
  • Total Volume Capacity (mL)3
  • ApplicationsExtraction of hydrophobic and polar analytes
  • SorbentC8
Key Features
  • Features C8 sorbent
  • Applications: Extraction of hydrophobic and polar analytes
More About this Item
TELOS nonpolar (reversed phase) SPE columns are available in a wide range of sorbents for extraction of hydrophobic and polar analytes. Select columns with endcaps for applications requiring minimal secondary silanol interactions to enhance compound retention and elution. All columns are made of polypropylene and feature Luer-slip outlet. TELOS C8 has a pore size of 70 Å and average particle size of 50 µm.