Cole-Parmer Short Thread Cap, 9 mm, Open Black Polypropylene, UltraBond Silicone/PTFE Septa; 1000/pk
Representative image only.
Kinesis Item # WZ-98703-63
No more damaged autosampler needles or sample loss
  • Ensure septa is not deformed due to overtightening—extends the life of your needle
  • Prevent sample evaporation due to poor vial-septa-cap sealing
  • UltraClean™ high-purity septa are optimized for GC and headspace analysis
  • Select caps with UltraBond® septum to avoid sample contamination
  • Silicone/PTFE septa offers the cleanest solution for contaminant free sample handling
  • UltraBond septa is bonded without adhesive, avoiding contamination of the sample
  • Designed with a black open cap with with silicone and red PTFE septa
PRODUCT OPTIONS
£443.32GBP / Pkg of 1000
£531.98GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Cap TypeShort Thread Screw
  • Cap MaterialPolypropylene
  • Cap colorBlack
  • Septum TypeUltraBond Silicone/PTFE
  • SterileNo
  • Description Short Thread Cap, 9 mm, Open Black Polypropylene, UltraBond Silicone/PTFE Septa; 1000/pk
Key Features
  • Ensure septa is not deformed due to overtightening—extends the life of your needle
  • Prevent sample evaporation due to poor vial-septa-cap sealing
  • UltraClean™ high-purity septa are optimized for GC and headspace analysis
  • Select caps with UltraBond® septum to avoid sample contamination
  • Silicone/PTFE septa offers the cleanest solution for contaminant free sample handling
  • UltraBond septa is bonded without adhesive, avoiding contamination of the sample
  • Designed with a black open cap with with silicone and red PTFE septa
More About this Item
Short thread polypropylene caps fit short thread vials. Caps are available in different colors, septum materials, and open or closed configurations to suit your application. UltraBond cap and septum form a one-piece unit using a bonding process without adhesives for contamination-free results. Pre-slit septa prevent vacuum forming inside the vial.