This item is now discontinued. Please browse similar items
Kinesis KX Membrane Filter, Hydrophilic PTFE, 47 mm dia., 0.45 µm; 100/pk
Representative image only.
Kinesis Item # WZ-12918-01
Remove particulate without adversely affecting your final product
  • Use membrane filtration for samples holding suspended solids
  • Highly efficient filtration of particles according to size
  • Complete sample filtration with the least sample loss and energy use
  • Filtration for HPLC sample preparation and mobile phase
  • Uniform manufacturing for repeatable results
SPECS & DESCRIPTION
Specifications & Description
  • MaterialHydrophilic PTFE
  • Pore Size (µm)0.45
  • Diameter (mm)47
  • DescriptionMembrane Filter, Hydrophilic PTFE, 47 mm dia., 0.45 µm; 100/pk
Key Features
  • Use membrane filtration for samples holding suspended solids
  • Highly efficient filtration of particles according to size
  • Complete sample filtration with the least sample loss and energy use
  • Filtration for HPLC sample preparation and mobile phase
  • Uniform manufacturing for repeatable results