Kinesis KX 96-Well Microplate, Deep Well, Square, Toughened PP, 2 mL; 50/PK

Representative image only.
Kinesis Item # WZ-12920-21
Don't compromise your research
  • RNase, DNase and endotoxin free for higher purity and better results
  • Polypropylene plates are compatible with solvents and additives
  • Withstand temperatures from -80 to 120°C, ideal for both molecular and storage applications
  • Deep well, square with V-shaped bottom, 2 mL/well, toughened PP

PRODUCT OPTIONS

£272.55GBP / Pkg of 50
£327.06GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • Number of Wells96
  • Well Volume (mL)2
  • Well ShapeSquare with V-shaped bottom
  • Plate TypeDeep Well
  • MaterialPolypropylene (PP)
  • Description96-Well Microplate, Deep Well, Square, Toughened PP, 2 mL; 50/PK

Key Features

  • RNase, DNase and endotoxin free for higher purity and better results
  • Polypropylene plates are compatible with solvents and additives
  • Withstand temperatures from -80 to 120°C, ideal for both molecular and storage applications
  • Deep well, square with V-shaped bottom, 2 mL/well, toughened PP

More About this Item

These 96-well microplates are manufactured from 100% virgin polypropylene (PP), with a toughened option available for long-term storage. Select round-bottom plates for more efficient resuspension or V-bottom plates for precise sample recovery. Plates are RNase, DNase and endotoxin-free to ensure purity and improve results of PCR and other molecular techniques. Use with microplate sealing mats to reduce evaporation and contamination.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat