Heraeus Noblelight Hollow Cathode Lamp, thallium, 50 mm, Perkin Elmer Lumina coded
Representative image only.
Heraeus Noblelight Item # WZ-11952-99
Achieve reliable atomic absorption results with high-performance lamps for your spectrometers
  • Carry out quicker start-up times
  • Reduce your operating costs with longer lifetime amp hours
  • Continuous light intensity for longer periods of time
  • Data-coded and uncoded versions are available
PRODUCT OPTIONS
£469.46GBP / Pkg of 1
£563.35GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialThallium
  • Compatible WithPerkin Elmer Lumina
  • Description Hollow Cathode Lamp, thallium, 50 mm, Perkin Elmer Lumina coded
Key Features
  • Carry out quicker start-up times
  • Reduce your operating costs with longer lifetime amp hours
  • Continuous light intensity for longer periods of time
  • Data-coded and uncoded versions are available
More About this Item
Heraeus Noblelight thallium (Tl) single-element hollow cathode lamps are quick to reach equilibrium performance, offer a long-term constant light intensity, and low noise with good calibration. Inside each lamp, there is a high volume of inert gas to increase the lamp’s lifetime to 5000 mA hours. Choose from coded lamp versions for Perkin Elmer®, Varian/Agilent, and Unicam systems.