Heraeus Noblelight Hollow Cathode Lamp, arsenic, 37 mm, Varian coded
Representative image only.
Heraeus Noblelight Item # WZ-11949-30
Achieve reliable atomic absorption results with high-performance lamps for your spectrometers
  • Carry out quicker start-up times
  • Reduce your operating costs with longer lifetime amp hours
  • Continuous light intensity for longer periods of time
  • Data-coded and uncoded versions are available
PRODUCT OPTIONS
£333.35GBP / Pkg of 1
£400.02GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialArsenic
  • Compatible WithVarian
  • Description Hollow Cathode Lamp, arsenic, 37 mm, Varian coded
Key Features
  • Carry out quicker start-up times
  • Reduce your operating costs with longer lifetime amp hours
  • Continuous light intensity for longer periods of time
  • Data-coded and uncoded versions are available
More About this Item
Heraeus Noblelight arsenic (As) single-element hollow cathode lamps are quick to reach equilibrium performance, offer a long-term constant light intensity, and low noise with good calibration. Inside each lamp, there is a high volume of inert gas to increase the lamp’s lifetime to 5000 mA hours. Choose from coded lamp versions for Perkin Elmer®, Varian/Agilent, and Unicam systems.