Kinesis Deuterium (D2) Detector Lamp for PE Lambda 2 to 45, 800, 900, Bio, 55X Series, LC480; 1/EA
Representative image only.
Kinesis Item # WZ-11941-24
Economical replacement deuterium lamps save you money
  • Select from standard or long life lamps
  • Manufactured in an ISO-accredited Facility
  • Individually Tested and Issued with a Test Certificate
  • Standard life 1000 hour bulb
  • 1 year warranty
PRODUCT OPTIONS
£478.35GBP / Each
£574.02GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithPerkin Elmer Lambda 2 to 45, 800, 900, Bio, 55X series, LC480
  • DescriptionD2 Detector Lamp for PE Lambda 2 to 45, 800, 900, Bio, 55X Series, LC480; 1/EA
  • Warranty1 year
1 Year Warranty
Key Features
  • Select from standard or long life lamps
  • Manufactured in an ISO-accredited Facility
  • Individually Tested and Issued with a Test Certificate
  • Standard life 1000 hour bulb
  • 1 year warranty
More About this Item
Our range of deuterium (D2) lamps are made to fit the majority of detectors used in analytical labs today. Our lamps are used by many of the major pharmaceutical and contract research organizations (CROs) around the globe.
Our lamps are manufactured under stringent ISO 9001 quality procedures. Each lamp is manufactured to rigorous standards and traceable from manufacturing to customer delivery.