This item is now discontinued. Please browse similar items
Kinesis Deuterium (D2) Detector Lamp for Jasco 970, 975 (B & C), 1570, 1575, 2070, 2075
Representative image only.
Kinesis Item # WZ-11941-58
Economical replacement deuterium lamps save you money
  • Select from standard or long life lamps
  • Manufactured in an ISO-accredited Facility
  • Individually Tested and Issued with a Test Certificate
  • Long life 2000 hour bulb
  • 6 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithJasco 970, 975 (B & C), 1570, 1575, 2070, 2075
  • DescriptionDeuterium (D2) Detector Lamp for Jasco 970, 975 (B & C), 1570, 1575, 2070, 2075
  • Warranty6 month
6 Month Warranty
Key Features
  • Select from standard or long life lamps
  • Manufactured in an ISO-accredited Facility
  • Individually Tested and Issued with a Test Certificate
  • Long life 2000 hour bulb
  • 6 month warranty
More About this Item
Our range of deuterium (D2) lamps are made to fit the majority of detectors used in analytical labs today. Our lamps are used by many of the major pharmaceutical and contract research organizations (CROs) around the globe.
Our lamps are manufactured under stringent ISO 9001 quality procedures. Each lamp is manufactured to rigorous standards and traceable from manufacturing to customer delivery.