This item is now discontinued. We recommend the following as suggested replacements: WZ-98704-81
Kinesis Gold Aluminum/Stainless Steel, Magnetic Crimp Cap, 20 mm Dia., Silicone/PTFE Septum w/8 mm Hole; 1000/pk
Representative image only.
Kinesis Item # WZ-98704-82
Aluminum crimps tightly around the neck for a tight seal
  • Select from a variety of caps and septa materials that fit your application and needs
  • Magnetic bimetallic caps available
  • Gold aluminum/stainless steel, magnetic 20 mm dia. cap Silicone blue/PTFE septum with an 8 mm hole Septum Hardness: 45° shore A Thickness: 3.2 mm
SPECS & DESCRIPTION
Specifications & Description
  • Cap TypeCrimp
  • Cap Diameter (mm)20
  • Cap MaterialMagnetic
  • Cap colorGold
  • Septum TypeBlue Silicone/PTFE
  • SterileNo
  • DescriptionCrimp Cap, 20mm, Magnetic Gold, 8mm Center Hole, Blue Silicone/PTFE Septa; 1000/pk
Key Features
  • Select from a variety of caps and septa materials that fit your application and needs
  • Magnetic bimetallic caps available
  • Gold aluminum/stainless steel, magnetic 20 mm dia. cap Silicone blue/PTFE septum with an 8 mm hole Septum Hardness: 45° shore A Thickness: 3.2 mm
More About this Item
Constructed of aluminum, these crimp vials are ideal for use with your crimp neck vials. To ensure autosamplers function reliably, these caps and septa meet the rigorous specifications demanded by both the analysis and the instrument itself. Defined hardness and cleanliness properties ensure there is a septum available to be both fully compatible with the autosampler needle and meet the demands of the specific analysis.