This item is now discontinued. Please browse similar items
Kinesis Red Aluminum/Stainless Steel Magnetic Crimp Cap, 20 mm Dia., Silicone/PTFE Septum w/8 mm Hole; 1000/pk
Representative image only.
Kinesis Item # WZ-98704-76
Aluminum crimps tightly around the neck for a tight seal
  • Select from a variety of caps and septa materials that fit your application and needs
  • Magnetic bimetallic caps available
  • Red aluminum/stainless steel, magnetic 20 mm dia., cap Silicone white/PTFE beige septum with an 8 mm hole Septum Hardness: 45° shore A Thickness: 3.0 mm
SPECS & DESCRIPTION
Specifications & Description
  • Cap TypeCrimp
  • Cap Diameter (mm)20
  • Cap MaterialMagnetic
  • Cap colorRed
  • Septum TypeSilicone/Beige PTFE
  • SterileNo
  • DescriptionCrimp Cap, 20mm, Magnetic Bimetallic Red, Silicone/Beige PTFE Septa; 1000/pk
Key Features
  • Select from a variety of caps and septa materials that fit your application and needs
  • Magnetic bimetallic caps available
  • Red aluminum/stainless steel, magnetic 20 mm dia., cap Silicone white/PTFE beige septum with an 8 mm hole Septum Hardness: 45° shore A Thickness: 3.0 mm
More About this Item
Constructed of aluminum, these crimp vials are ideal for use with your crimp neck vials. To ensure autosamplers function reliably, these caps and septa meet the rigorous specifications demanded by both the analysis and the instrument itself. Defined hardness and cleanliness properties ensure there is a septum available to be both fully compatible with the autosampler needle and meet the demands of the specific analysis.