This item is now discontinued. Please browse similar items
DWK Life Sciences (Kimble) 747010-0000 Microscale Organic Chemistry Kit, threaded connections
Representative image only.
DWK Life Sciences (Kimble) Mfr # 747010-0000 – Item # WZ-34557-99
Kit is complete to handle most organic chemistry experiments!
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass
  • DescriptionMicroscale Organic Chemistry Kit, threaded connections
More About this Item
Organic Chemistry kit features threaded chaste connections that can be assembled "rightside up" or "upside down". High temperature resistant blue nylon connectors are ideal to use up to 200 °C. Kits are complete with a 13-1/2" x 10 x 2-1/2" rugged polyethylene case with die-cut foam insert.