Kap-Uts 2007-25002 Polypropylene Culture Tube Closures, 25 mm, red
Representative image only.
Kap-Uts Mfr # 2007-25002 – Item # WZ-04501-06
  • Closures minimize evaporation while allowing necessary gas exchange.
  • Positive-grip closures of stainless steel or autoclavable polypropylene.
PRODUCT OPTIONS
£228.01GBP / Case of 500
£273.61GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionPolypropylene Culture Tube Closures, 25 mm, red
Key Features
  • Closures minimize evaporation while allowing necessary gas exchange.
  • Positive-grip closures of stainless steel or autoclavable polypropylene.
More About this Item
These new tube closures ensure the viability of microbiological cultures by protecting solid and broth media from excessive evaporation. They fit snugly on rimless culture tubes and flasks while allowing for gas exchange and ventilation.