This item is now discontinued. Please browse similar items
Kanomax Airflow Transducer Probe, Omni, Spherical, 16.125" L, 50 m/s max; No Cable
Representative image only.
Kanomax Mfr # 0965-10 – Item # WZ-10006-69
Work with the Kanomax airflow transducers
  • Built-in temperature compensation sensor in the probe provides more accurate measurements
  • Smart probe technology lets you change probes without recalibrating the transducer
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Probe length (in)16.125
  • DescriptionAirflow Transducer Probe, Omni, Spherical, 16.125\" L, 50 m/s max; No Cable
  • Warranty1 year
1 Year Warranty
Key Features
  • Built-in temperature compensation sensor in the probe provides more accurate measurements
  • Smart probe technology lets you change probes without recalibrating the transducer