Justrite Quick-Disconnect Disposal Can; 1 Gal
Representative image only.
Justrite Mfr # 12160 – Item # WZ-12021-08
Remove can and empty waste without disconnecting your inlet tubing
  • Safe method to stage and draw reagents
  • FM approved
  • Quick disconnects allow simple attachment of one inlet and one vent
  • Eliminate safety concerns and accidents associated with glass bottles
  • Polypropylene/EPDM quick-disconnect fittings features 3/8" drain orifice
PRODUCT OPTIONS
£219.18GBP / Each
£263.02GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (Gal)1
  • Capacity (Liters)4
  • Exterior Height (in)12.75
  • Exterior Diameter (in)7.75
  • Exterior Height (cm)32.4
  • Exterior Diameter (cm)19.7
  • ApprovalFM
  • DescriptionQuick-Disconnect Disposal Can; 1 Gal
Key Features
  • Safe method to stage and draw reagents
  • FM approved
  • Quick disconnects allow simple attachment of one inlet and one vent
  • Eliminate safety concerns and accidents associated with glass bottles
  • Polypropylene/EPDM quick-disconnect fittings features 3/8" drain orifice
More About this Item
Polyethylene disposal cans provide a safe hands-free way to drain waste from your HPLC application. A spring-loaded closure opens when contents are subjected to pressure, being FM approved. Double fittings eliminate back pressure and help control vapors. The quick disconnect mechanisms allow you to remove and empty the waste without having to disconnect tubing from your application.
An optional spill basin captures any leakage and provides additional protection against accidental pumping.