Justrite 28178 HPLC Poly Manifold w/Polypropylene Quick Disconnect for Safety Disposal Can
Representative image only.
Justrite Mfr # 28178 – Item # WZ-86394-29
A cost-effective way to collect HPLC waste
PRODUCT OPTIONS
£226.28GBP / Each
£271.54GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene(PP)
  • DescriptionHPLC Poly Manifold w/polypropylene quick disconnect
More About this Item
These polypropylene manifolds accept up to six waste lines from HPLC processes to be fed into a single intake fitting. Manifolds have six 1/4-28 ports, one 3/8" NPT port (all come with caps for unused ports), and one quick-disconnect port in either 316 stainless steel or polypropylene.