Justrite 28177 HPLC Poly Manifold w/Stainless Steel Quick Disconnect for Safety Disposal Can
Representative image only.
Justrite Mfr # 28177 – Item # WZ-86394-27
A cost-effective way to collect HPLC waste
PRODUCT OPTIONS
£299.17GBP / Each
£359.00GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene(PP)
  • DescriptionHPLC Poly Manifold w/stainless quick disconnect
More About this Item
These polypropylene manifolds accept up to six waste lines from HPLC processes to be fed into a single intake fitting. Manifolds have six 1/4-28 ports, one 3/8" NPT port (all come with caps for unused ports), and one quick-disconnect port in either 316 stainless steel or polypropylene.