This item is now discontinued. Please browse similar items
Justrite 26753 Drum Cover with Vent and Gasket for 55 Gal Drum, Steel, Self-Latching
Representative image only.
Justrite Mfr # 26753 – Item # WZ-81792-54
Easily convert your open-top drums into fire-safe receptacles
  • Spring-loaded cover securely closes via gravity
  • TÜV certified
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (Liters)208.2
  • Capacity (Gal)55
  • ColorRed
  • Self-ClosingYes
  • ComplianceFM, NESAM
  • Shipping Weight (lbs)15
  • DescriptionDrum cover for 55 gal. drum, self-latching with vent and gasket
  • Warranty1 Year
1 Year Warranty
Key Features
  • Spring-loaded cover securely closes via gravity
  • TÜV certified
  • 1 year warranty
More About this Item
These red drum covers allow you to convert steel drums into safe waste receptacles. Keep facilities protected against fires from combustible trash. Features a spring-loaded, rotary latch mechanism, which allows the cover to securely close via gravity or a slight touch of the hand providing an airtight seal. This reduces the potential for the drum lid to be left open by mistake, eliminating the chance for oxygen to enter and start a fire.
Covers are manufactured of steel, with a highly chemical resistant powder-coat finish. You only need a screwdriver and wrench to attachthe cover and it can be quickly removed for emptying the drum contents.