Justrite 12754 Liquid Disposal Safety Can, Polyethylene, 5 Gal
Representative image only.
Justrite Mfr # 12754 – Item # WZ-86395-24
Allows quick visual inspection of liquid level
  • Flame arrester guards against outside ignition
  • Automatically vents at between 3 and 5 psig
  • OSHA and NFPA compliant
  • FM Approved
  • 10 year warranty
PRODUCT OPTIONS
£388.38GBP / Each
£466.06GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialHigh Density Polyethyene (HDPE)
  • Capacity (Gal)5
  • Capacity (Liters)18.93
  • Exterior Height (in)20
  • Exterior Diameter (in)12
  • Exterior Height (cm)50.8
  • Exterior Diameter (cm)30.48
  • ApprovalFM
  • DescriptionTranslucent Liquid Disposal Can, 5 Gal.
  • Warranty10 Years
10 Year Warranty
Key Features
  • Flame arrester guards against outside ignition
  • Automatically vents at between 3 and 5 psig
  • OSHA and NFPA compliant
  • FM Approved
  • 10 year warranty
More About this Item
These liquid disposal safety cans offer you a better solution for the containment of flammable liquid or flammable liquid waste. Constructed of durable translucent polyethylene makes it easily to detect liquid level. Large opening with cap held open provides convenient filling and pouring. The cans also offer superior chemical resistance and leak protection.
Conductive, patented carbon insert embedded in the container allows for proper grounding. Broad-base design provides stability and resists accidental tip over. Self-closing lid automatically vents between 3 and 5 psi (0.2 and 0.35 bar). Built-in stainless steel flame arrester guards against inside vapor ignition. Stainless steel hardware is corrosive resistant.