This item is now discontinued. Please browse similar items
Julabo SW22 and 23 Lift-up Stainless Steel Bath Cover
Representative image only.
Julabo Mfr # 8970268 – Item # WZ-12149-51
Quality accessories to add to your shaker bath
  • Choose from lids, racks and trays for your Julabo shaker
  • Bath cover is ideal up to temperatures up to 100 deg C.
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Temperature (° C)100
  • Working Area Width (in)19 3/4
  • Working Area Length (in)11 7/8
  • Wetted MaterialsMakrolon
  • DescriptionLift-up Stainless Steel Bath Cover
Two year warrantyConformite Europeenne
Key Features
  • Choose from lids, racks and trays for your Julabo shaker
  • Bath cover is ideal up to temperatures up to 100 deg C.
  • 2 year warranty
More About this Item
Lift up cover prevents liquid loss due to evaporation, and protects from contamination. Stainless steel cover fits Julabo SW22 and 23 Shaking Water Baths (12149-00, 12149-05, 12149-10, and 12149-15)