Jenway 924 001 General Purpose Epoxy Bodied pH Electrode
Representative image only.
Jenway Item # WZ-99960-34
A variety of pH electrodes for your Jewnay meters
  • Combination electrodes
  • Application specific electrodes
  • Waterproof electrodes
  • Temperature and reference electrodes
PRODUCT OPTIONS
£79.04GBP / Each
£94.85GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsStandard measurements of general solutions
  • Electrode TypeCombination
  • Electrode Body MaterialEpoxy
  • Reference JunctionPorous PTFE
  • Reference ElectrolyteAgCl, gel filled single junction
  • Min pH (pH)0
  • Max pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • ConnectorBNC
  • Diameter (mm)12
  • Length (mm)120
  • Cable Length (m)1
  • DescriptionGeneral Purpose Epoxy Bodied pH Electrode
Key Features
  • Combination electrodes
  • Application specific electrodes
  • Waterproof electrodes
  • Temperature and reference electrodes
More About this Item
Choose the right electrode for your application.