This item is now discontinued. Please browse similar items
Jenway 740701 Combined Sipper Pump and Peltier Cell
Representative image only.
Jenway Item # WZ-83056-43
Optimize performance of your Jenway® spectrophotometer
  • Expand analysis capabilities or make testing easier
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithJenway 7410, 7415, and 7615 Spectrophotometers
  • DescriptionCombined Sipper Pump and Peltier Cell
  • Warranty1 year
1 Year Warranty
Key Features
  • Expand analysis capabilities or make testing easier
  • 1 year warranty
More About this Item
Choose from a wide range of accessories including communication cables, replacement bulbs, and more for your Jenway® spectrophotomer.
For safe and accurate sample handling the sipper pump will circulate samples and can be programmed to deliver reproducible sample volumes. Choose from manual or timed mode to accommodate any application. For applications where sample temperature is critical, the Peltier accessory is offered which enables sample temperature to be set and controlled. The current temperature is displayed above the set point temperature on the LCD, and can be set in °C or °F. The combined sipper-Peltier pump combines the functionality of both the sipper pump and the Peltier enabling sample temperature to be controlled while the sample is being circulated.